Learn More
Insulin Receptor (α-subunit) Ab-2 Mouse Monoclonal Antibody, Epredia™
Epredia™ Insulin Receptor (α-Subunit) Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.
Marke: Epredia MS-632-PABX
Weitere Details : Weight : 1.00000kg
Beschreibung
The human insulin receptor is a heterotetrameric membrane glycoprotein consisting of disulfide-linked subunits in a Β-α-α-Β configuration. The Β-subunit (95kDa) possesses a single transmembrane domain, whereas the α-subunit (135kDa) is completely extracellular.Host Species: Mouse
Clone: 83-7
Isotype: IgG1/κ
Species Reactivity: Human, Cow, Pig, Sheep and Rabbit. Does not react with mouse and rat.
Epitope: Exon 3
Immunogen: IM-9 lymphocytes followed by IR
Molecular Weight: 135kDa
Positive Control: IM-9 lymphocyte cells, Placenta
Cellular Localization: Cell membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- ELISA
- Flow Cytometry
- Insulin-like Agonist
- Does Not Significantly Inhibit Insulin Binding
- Tyrosine Kinase Assay (Ab-mediated capture on microplates)
Note:
USA: RUO; Int'l: RUO
Spezifikation
Insulin Receptor (alpha-Subunit) Ab-2 | |
Monoclonal | |
1mg/mL | |
Mouse | |
200 μL | |
Steroids Hormones and Growth Factors | |
Bovine, Human, Porcine, Rabbit, Sheep | |
IgG2a κ |
Immunohistochemistry (Paraffin) | |
83-7 | |
Unconjugated | |
IM-9 lymphocytes followed by IR | |
RUO | |
Primary | |
Concentrated |