missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ S100 A6 Ab-9 Rabbit Polyclonal Antibody with BSA and azide

Epredia™ S100 A6 Ab-9, Rabbit Polyclonal Antibody provides accurate, reproducible results.

Brand:  Epredia™ RB-1805-A1

 View more versions of this product

Product Code. 12653157

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

This item is not returnable. View return policy

Description

Description

S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an α and β chain whereas S100B is composed of two β chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat, Cow, Horse, Cat and Dog. Others not known.

Epitope: Not determined

Immunogen: Purified recombinant human S100 A6 monomer expressed in E. coli

Molecular Weight: 10-12kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

S100 A6 Ab-9
Polyclonal
Rabbit
Tonsil
RUO
Primary
Concentrated
Immunohistochemistry (Paraffin)
Unconjugated
Purified recombinant human S100 A6 monomer expressed in E. coli
500 μL
Neuroscience
Bovine, Canine, Equine, Feline, Human, Mouse, Rat
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.