Learn More
Epredia™ Lab Vision™ S100 A6 Ab-9 Rabbit Polyclonal Antibody with BSA and azide
Epredia™ S100 A6 Ab-9, Rabbit Polyclonal Antibody provides accurate, reproducible results.
137.00€ - 765.00€
Specifications
| Antigen | S100 A6 Ab-9 |
|---|---|
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|
12633157
|
Epredia™
RB-1805-A0 |
100 μL | Concentrated | ||||||
|
12653157
|
Epredia™
RB-1805-A1 |
500 μL | Concentrated | ||||||
|
12623157
|
Epredia™
RB-1805-A |
1 mL | Concentrated | ||||||
|
12673157
|
Epredia™
RB-1805-R7 |
7 mL | Prediluted | ||||||
Description
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an α and β chain whereas S100B is composed of two β chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse, Rat, Cow, Horse, Cat and Dog. Others not known.
Epitope: Not determined
Immunogen: Purified recombinant human S100 A6 monomer expressed in E. coli
Molecular Weight: 10-12kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
| S100 A6 Ab-9 | |
| Polyclonal | |
| Rabbit | |
| Neuroscience | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Purified recombinant human S100 A6 monomer expressed in E. coli | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.