missing translation for 'onlineSavingsMsg'
Learn More

LAMP-1/CD107a Antibody (5H6), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication

Brand:  Novus Biologicals NBP2-25154SS

 View more versions of this product

Product Code. 18088885

  • 200.00€ / 25µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LAMP-1/CD107a Monoclonal specifically detects LAMP-1/CD107a in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LAMP-1/CD107a
Monoclonal
1 mg/mL
Western Blot 1:1000-1:10000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:2000
P11279
LAMP1
Amino acids 32-350 of the human LAMP1 precursor sequence. [UniProt# P11279]
Affinity Purified
RUO
Primary
Human, Bovine, Porcine, Mouse and Rat. Other species have not been tested.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
5H6
Unconjugated
PBS, 50% glycerol with 5mM Sodium Azide
CD107 antigen-like family member A, CD107a, CD107a antigen, LAMP-1, LAMPA, LGP120, lysosomal-associated membrane protein 1, lysosome-associated membrane glycoprotein 1, Lysosome-associated membrane protein 1
Mouse
90-120 kDa
0.025 mL
Autophagy, Cholesterol Metabolism, Endosome Markers, Glycobiology, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones, Virology Bacteria and Parasites
3916
Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only