missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMP-1/CD107a Antibody (5H6), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
200.00€ - 558.00€
Specifications
| Antigen | LAMP-1/CD107a |
|---|---|
| Clone | 5H6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000-1:10000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:2000 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18088885
|
Novus Biologicals
NBP2-25154SS |
0.025 mL |
200.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18756003
|
Novus Biologicals
NBP2-25154 |
558.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
LAMP-1/CD107a Monoclonal specifically detects LAMP-1/CD107a in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| LAMP-1/CD107a | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| P11279 | |
| 3916 | |
| Amino acids 32-350 of the human LAMP1 precursor sequence. [UniProt# P11279] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 90-120 kDa |
| 5H6 | |
| Western Blot 1:1000-1:10000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:2000 | |
| Monoclonal | |
| Mouse | |
| Autophagy, Cholesterol Metabolism, Endosome Markers, Glycobiology, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones, Virology Bacteria and Parasites | |
| PBS, 50% glycerol with 5mM Sodium Azide | |
| CD107 antigen-like family member A, CD107a, CD107a antigen, LAMP-1, LAMPA, LGP120, lysosomal-associated membrane protein 1, lysosome-associated membrane glycoprotein 1, Lysosome-associated membrane protein 1 | |
| LAMP1 | |
| IgG1 | |
| Affinity Purified | |
| Human, Bovine, Porcine, Mouse and Rat. Other species have not been tested. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title