missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ Tb-pERM (pLRRKtide) Antibody

A phospho specific antibody directed to the ezrin/radixin/moesin (ERM) peptide that is a substrate for LRRK2 (Leucine-rich repeat kinase 2) and LRRK2 mutants.

Brand:  Invitrogen™ PV4897

 View more versions of this product

Product Code. 10402175

  • 4475.00€ / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

A phospho specific antibody directed to the ezrin/radixin/moesin (ERM) peptide that is a substrate for LRRK2 (Leucine-rich repeat kinase 2) and LRRK2 mutants. Mutations in LRRK2 have been found to be related to late-onset Parkinson's disease.

For use in combination with the Fluorescein-ERM (LRRKtide) substrate (Cat. No. PV4901) to evaluate inhibitors in medium to high throughput screening applications.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Serine/threonine kinase
0.52mg/mL
150kDa
RUO
Primary
Terbium-labeled pERM (pLRRKtide) antibody in HEPES buffered saline. Store at -20°C.
340/490, 520 nm
Dry Ice
Hu
LanthaScreen
Kinase Assay
Tb (Terbium)/Lanthanide Chelates
25 μg
Target and Lead Identification and Validation
1 Each
Liquid
Human
End Point
Ser/Thr Kinases
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.