Learn More
Invitrogen™ LanthaScreen™ Tb-pERM (pLRRKtide) Antibody
A phospho specific antibody directed to the ezrin/radixin/moesin (ERM) peptide that is a substrate for LRRK2 (Leucine-rich repeat kinase 2) and LRRK2 mutants.
4475.00€ - 21840.00€
Specifications
Excitation/Emission 340/490, 520 nm
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10402175
View Documents
Invitrogen™
PV4897
25 μg
4475.00€
25µg
View alternative products
Save Up to 
»
10749135
View Documents
Invitrogen™
PV4898
1 mg
21840.00€
1mg
View alternative products
Save Up to 
»
Description
Description
A phospho specific antibody directed to the ezrin/radixin/moesin (ERM) peptide that is a substrate for LRRK2 (Leucine-rich repeat kinase 2) and LRRK2 mutants. Mutations in LRRK2 have been found to be related to late-onset Parkinson's disease.
For use in combination with the Fluorescein-ERM (LRRKtide) substrate (Cat. No. PV4901) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
Specifications
340/490, 520 nm