Description
The LC3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human LC3. This antibody recognizes human, mouse, rat antigen. The LC3 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™488.
Specifications
Specifications
| Antigen | LC3 |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Polyclonal |
| Conjugate | CoraLite 488 |
| Host Species | Rabbit |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 81631 |
| Target Species | Human, Rat, Mouse |
| Isotype | IgG |