missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LDB3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17732504
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17732504 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17732504 Supplier Invitrogen™ Supplier No. PA5121105

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HL-60, MCF7, U-937, Raji, BT-474, Mouse skeletal muscle, Rat skeletal muscle. The target is usually found in the following locations: Cell projection, Cytoplasm, Z line, cytoskeleton, myofibril, perinuclear region, pseudopodium, sarcomere. Immunogen sequence: MSYSVTLTGP GPWGFRLQGG KDFNMPLTIS RITPGSKAAQ SQLSQGDLVV AIDGVNTDTM THLEAQNKIK SASYNLSLTL QKSKRPIPIS TTAPPVQTPL PVIPHQKVVV NSPANADYQE RFNPSALKDS ALSTHKPIEV KGLGGKATII HAQYNTPISM YSQDAIMDAI AGQAQAQGSD FSGSLPIKDL AVDSASPVYQ AVIKSQNKPE DEADEWARRS SNLQSRSFRI LAQMTGTEFM QDPDEEALRR SRERFETERN SPRFAKLRNW HHGLSAQILN VKS.

This gene encodes a PDZ domain-containing protein. PDZ motifs are modular protein-protein interaction domains consisting of 80-120 amino acid residues. PDZ domain-containing proteins interact with each other in cytoskeletal assembly or with other proteins involved in targeting and clustering of membrane proteins. The protein encoded by this gene interacts with alpha-actinin-2 through its N-terminal PDZ domain and with protein kinase C via its C-terminal LIM domains. The LIM domain is a cysteine-rich motif defined by 50-60 amino acids containing two zinc-binding modules. This protein also interacts with all three members of the myozenin family. Mutations in this gene have been associated with myofibrillar myopathy and dilated cardiomyopathy. Alternatively spliced transcript variants encoding different isoforms have been identified; all isoforms have N-terminal PDZ domains while only longer isoforms (1, 2 and 5) have C-terminal LIM domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDB3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LDB3
Gene Accession No. O75112, Q9JKS4
Gene Alias AW742271; cardiomyopathy, dilated 1C (autosomal dominant); CMD1C; CMH24; CMPD3; cypher; FLJ35865; HGNC:15710; KIAA01613; KIAA0613; LDB3; LDB3Z1; LDB3Z4; LIM domain binding 3; LIM domain-binding protein 3; LVNC3; MFM4; ORACLE; oracle protein; PDLIM6; PDZ and LIM domain 6; PDZ-LIM domain protein; protein cypher; Protein oracle; RGD1564875; ZASP; Z-band alternatively spliced PDZ motif protein; Z-band alternatively spliced PDZ-motif protein
Gene Symbols LDB3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-283 of human LDB3 (NP_001073585.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11155, 24131, 498587
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.