missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) Perchlorate Trihydrate, ACS Reagent, 98%, Honeywell Fluka™

Product Code. 15645290 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15645290 100 g Glass Bottle 100g
15655290 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15645290 Supplier Honeywell-Fluka Supplier No. 383066100G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent, 98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13453-62-8
Molecular Formula Cl2H6O11Pb
Molecular Weight (g/mol) 460.10
MDL Number MFCD00149817
InChI Key KKGGEAQRICYXNM-UHFFFAOYSA-L
Synonym lead ii perchlorate trihydrate, acmc-20aknd, 2clo4.pb.3h2o, lead perchlorate 3h2o, lead ii perchlorate, trihydrate, lead perchlorate, trihydrate, reagent acs, lead ii perchlorate trihydrate, acs reagent
PubChem CID 25021834
IUPAC Name λ²-lead(2+) trihydrate diperchlorate
SMILES O.O.O.[Pb++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Melting Point 83°C to 84°C
Color White
pH 3 to 5 (at 25°C)
Quantity 100 g
Linear Formula Pb(ClO4)2·3H2O
UN Number UN1470
Formula Weight 460.15g/mol
Percent Purity 98%
Packaging Glass Bottle
Appearance Crystalline
Density 2.6g/cm3
Chemical Name or Material Lead(II) perchlorate trihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.