missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEM3 Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-41144-0.025mg
This item is not returnable.
View return policy
Description
LEM3 Polyclonal specifically detects LEM3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| LEM3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| ANKLE1 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human ANKLE1. The immunogen is located within the last 50 amino acids of ANKLE1. Amino Acid Squence: CAGWPPARRRRLGVHL | |
| 0.025 mg | |
| Protein Kinase | |
| 126549 | |
| Human, Mouse, Rat | |
| IgY |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA | |
| ANKRD41, ankyrin repeat and LEM domain containing 1, ankyrin repeat and LEM domain-containing protein 1, FLJ39369, LEM domain containing 6, LEMD6, LEM-domain containing protein 3 | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Two isoforms of ANKLE1 are known to exist. ANKLE1 antibody is predicted to not cross-react with ANKLE2 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction