missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEM3 Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
168.00€ - 507.00€
Specifications
| Antigen | LEM3 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18602956
|
Novus Biologicals
NBP2-41144-0.025mg |
0.025 mg |
168.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18149927
|
Novus Biologicals
NBP2-41144 |
0.1 mg |
507.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LEM3 Polyclonal specifically detects LEM3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| LEM3 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| ANKRD41, ankyrin repeat and LEM domain containing 1, ankyrin repeat and LEM domain-containing protein 1, FLJ39369, LEM domain containing 6, LEMD6, LEM-domain containing protein 3 | |
| ANKLE1 | |
| IgY | |
| Affinity Purified |
| 1 mg/mL | |
| Polyclonal | |
| Chicken | |
| Protein Kinase | |
| PBS with 0.02% Sodium Azide | |
| 126549 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human ANKLE1. The immunogen is located within the last 50 amino acids of ANKLE1. Amino Acid Squence: CAGWPPARRRRLGVHL | |
| Primary | |
| Two isoforms of ANKLE1 are known to exist. ANKLE1 antibody is predicted to not cross-react with ANKLE2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title