missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leptin R Antibody (RM0085-9C29) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-21725-0.025MG
This item is not returnable.
View return policy
Description
Leptin R Monoclonal specifically detects Leptin R in Mouse samples. It is validated for Western Blot.
Specifications
| Leptin R | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1:1000 | |
| P48356 | |
| LEPR | |
| Mouse recombinant EC-domain of LeptinR | |
| 0.025 mg | |
| Signal Transduction | |
| 3953 | |
| Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot | |
| RM0085-9C29 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| CD295 antigen, DB, DKFZp686B1731, huB219, LEP-R, leptin receptor, OB receptor, OB-R, OBRCD295 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of Extracellular domain of Leptin R. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction