missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leptin R Antibody (RM0085-9C29) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
195.00€ - 523.00€
Specifications
| Antigen | Leptin R |
|---|---|
| Clone | RM0085-9C29 |
| Concentration | LYOPH |
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18231855
|
Novus Biologicals
NBP1-21725-0.025MG |
0.025 mg |
195.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18023024
|
Novus Biologicals
NBP1-21725 |
0.1 mg |
523.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Leptin R Monoclonal specifically detects Leptin R in Mouse samples. It is validated for Western Blot.Specifications
| Leptin R | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Signal Transduction | |
| P48356 | |
| 3953 | |
| Mouse recombinant EC-domain of LeptinR | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| RM0085-9C29 | |
| Western Blot 1:500-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CD295 antigen, DB, DKFZp686B1731, huB219, LEP-R, leptin receptor, OB receptor, OB-R, OBRCD295 | |
| LEPR | |
| Primary | |
| This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of Extracellular domain of Leptin R. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title