missing translation for 'onlineSavingsMsg'
Learn More

LGALS3BP Mouse anti-Human, CoraLite 594, Clone: 2D8E11, Proteintech

Product Code. 17731409 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17731409 20 μL 20µL
17721409 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17731409 Supplier Proteintech Supplier No. CL5946006620UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LGALS3BP Monoclonal antibody specifically detects LGALS3BP in Human samples. It is validated for IF

Specifications

Antigen LGALS3BP
Applications Immunofluorescence
Classification Monoclonal
Clone 2D8E11
Concentration 2000 μg/mL
Conjugate CoraLite 594
Formulation PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
Gene Accession No. BC015610
Host Species Mouse
Purification Method Protein A purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 3959
Target Species Human
Content And Storage Store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.