missing translation for 'onlineSavingsMsg'
Learn More

Limetol, TRC

Product Code. 30473630
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473630 100 mg 100mg
30483702 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30473630 Supplier Toronto Research Chemicals Supplier No. TRCL461520100MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7392-19-0
Molecular Formula C10 H18 O
Molecular Weight (g/mol) 154.25
Synonym 2H-Pyran, 2-ethenyltetrahydro-2,6,6-trimethyl-, 2H-Pyran, tetrahydro-2,2,6-trimethyl-6-vinyl- (8CI), Pyran, tetrahydro-2,2,6-trimethyl-6-vinyl- (7CI), 2-Ethenyltetrahydro-2,6,6-trimethyl-2H-pyran, 2,2,6-Trimethyl-6-ethenyltetrahydrofuran, 2,2,6-Trimethyl-6-vinyltetrahydropyran, 2,2,6-Trimethyl-6-vinyltetrahydropyrane, 2,6,6-Trimethyl-2-vinyltetrahydro-2H-pyran, 2,6,6-Trimethyl-2-vinyltetrahydropyran, Bois de Rose oxide, Limetol
IUPAC Name 2-ethenyl-2,6,6-trimethyloxane
SMILES CC1(C)CCCC(C)(O1)C=C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H18O/c1-5-10(4)8-6-7-9(2,3)11-10/h5H,1,6-8H2,2-4H3
Formula Weight 154.1358
Chemical Name or Material Limetol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.