missing translation for 'onlineSavingsMsg'
Learn More

Lithium perchlorate trihydrate, ACS, 63.0-68.0% LiClO{4}

Product Code. p-7080018
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15457125 10 g 10g
15467125 100 g 100g
15477125 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15457125 Supplier Thermo Scientific Alfa Aesar Supplier No. 045923.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13453-78-6 | ClH6LiO7 | 160.431 g/mol

Lithium perchlorate trihydrate is used as an oxygen source for chemical oxygen generators as it liberates oxygen at high temperatures. Excellent solubility of lithium perchlorate permits its use in Diels-Alder reactions. It is also employed in lithium-ion batteries as an electrolyte and acts as a chaotropic agent to denature proteins. It is used in the Baylis-Hillman reaction as a catalyst in the coupling of alpha, beta-unsaturated carbonyl compounds with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13453-78-6
Molecular Formula ClH6LiO7
Molecular Weight (g/mol) 160.431
MDL Number MFCD00149765
InChI Key JAWYRNYHJJDXHX-UHFFFAOYSA-M
Synonym lithium perchlorate trihydrate, perchloric acid, lithium salt, trihydrate, unii-jyk745743v, acmc-1c1zx, lithium perchlorate hydrate, lithiumperchlorate trihydrate, ksc491a4p, lithium pechlorate trihydrate, lithiumperchloratetrihydrate, lithotab trihydrate perchlorate ion
PubChem CID 23708903
IUPAC Name lithium;perchlorate;trihydrate
SMILES [Li+].O.O.O.[O-]Cl(=O)(=O)=O

Specifications

Color White
Physical Form Crystalline
Quantity 10 g
Linear Formula LiClO4·3H2O
UN Number UN1481
Merck Index 14,5539
Formula Weight 160.45 (106.40 Anhydrous)
Grade ACS
Concentration or Composition (by Analyte or Components) 63.0 to 68.0% LiClO4
Odor Odorless
Density 1.84g/mL
Chemical Name or Material Lithium perchlorate trihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.