missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPHN2 Polyclonal Antibody, Invitrogen™
Description
Immunogen sequence: PVKPVIGGSSS EDDAIVADAS SLMHSDNPGL ELHHKELEAP LIPQRTHSLL YQPQKKVKSE GTDSYVSQLT AEAEDHLQS Highest antigen sequence identity to the following orthologs - mouse 93%, rat 91%.
Calcium-independent receptor of low affinity for alpha-latrotoxin, an excitatory neurotoxin present in black widow spider venom which triggers massive exocytosis from neurons and neuroendocrine cells. Receptor propably implicated in the regulation of exocytosis.
Specifications
Specifications
| Antigen | LPHN2 |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ADGRL2 |
| Gene Accession No. | O95490 |
| Gene Alias | LPHN2; Cl2aa; CIRL2; CL2; Latrophilin 1; Latrophilin-2; Lectomedin-1; Latrophilin 2; Latrophilin homolog 2 (cow); KIAA0786; Beta-latrotoxin receptor; CIRL-2; Latrophilin homolog 1; Latrotoxin receptor 2; Latrotoxin receptor homolog 2; LEC1; LPHH1 |
| Gene Symbols | ADGRL2 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?