missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRK2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NB110-58771SS
This item is not returnable.
View return policy
Description
LRRK2 Polyclonal specifically detects LRRK2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Knockout Validated, Knockdown Validated.
Specifications
| LRRK2 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.05% Sodium Azide | |
| augmented in rheumatoid arthritis 17, AURA17, Dardarin, DKFZp434H2111, EC 2.7.11.1, FLJ45829, leucine-rich repeat kinase 2, leucine-rich repeat serine/threonine-protein kinase 2, PARK8DARDARIN, Parkinson disease (autosomal dominant) 8, RIPK7, ROCO2 | |
| Rabbit | |
| 286 kDa | |
| 0.025 mL | |
| Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| 120892 | |
| Human, Mouse, Rat, Primate | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow Cytometry, Immunoassay | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 24312256), Immunohistochemistry-Frozen, Flow (Cell Surface) reported in scientific literature (PMID 20483355), Knockout Validated, KnockDown Validated | |
| Q5S007 | |
| LRRK2 | |
| A synthetic peptide made to a C-terminal region, within residues 2500-2527 of the human LRRK2 protein, conjugated to KLH. [Swiss-Prot# Q5S007] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, rat, mouse and primate. Predicted to react with porcine based on 100% sequence homology. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction