missing translation for 'onlineSavingsMsg'
Learn More

Lynestrenol, TRC

Product Code. 30132778
Change view
Click to view available options
Quantity:
5 mg
10 mg
2.5 g
Unit Size:
10mg
2.5g
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132778 2.5 g 2.5g
30137125 5 mg 5mg
30003409 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132778 Supplier Toronto Research Chemicals Supplier No. TRCL4878502.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-76-6
Molecular Formula C20 H28 O
Molecular Weight (g/mol) 284.44
Synonym Lynestrenol, 19-Nor-17α-pregn-4-en-20-yn-17-ol, 19-Norpregn-4-en-20-yn-17-ol, (17α)-, 19-Nor-17α-pregn-4-en-20-yn-17-ol (6CI,8CI), 17-Ethynylestr-4-en-17β-ol, 17α-Ethinyl-17β-hydroxy-δ4-estrene, 17α-Ethynyl-17β-hydroxy-δ4-estrene, 17α-Ethynyl-17β-hydroxyestr-4-ene, 17α-Ethynyl-19-nor-4-androsten-17β-ol, 17α-Ethynylestr-4-en-17β-ol, 17α-Ethynylestrenol, 3-Desoxy-17α-ethinyl-19-nortestosterone, 3-Desoxynorlutin, Ethynylestrenol, Exlutena, Exluton, Exlutona, Fysionorm, Linestrenol, Lynestrenol, NSC 37725, NSC 73879, Org 485-50, Orgametil, Orgametril, Orgametrol, Ovamezzo, Ovoresta, Ovoresta M, Physiostat, δ4-17α-Ethinylestren-17β-ol
IUPAC Name (8R,9S,10R,13S,14S,17R)-17-ethynyl-13-methyl-2,3,6,7,8,9,10,11,12,14,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-17-ol
SMILES C[C@]12CC[C@H]3[C@@H](CCC4=CCCC[C@H]34)[C@@H]1CC[C@@]2(O)C#C

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C20H28O/c1-3-20(21)13-11-18-17-9-8-14-6-4-5-7-15(14)16(17)10-12-19(18,20)2/h1,6,15-18,21H,4-5,7-13H2,2H3/t15-,16+,17+,18-,19-,20-/m0/s1
Formula Weight 284.214
Chemical Name or Material Lynestrenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.