missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: QLYRYLLRCC QQLPTKGIQQ HYKHAVRQSF RVHSDEDNPE RIQQIIKRAI EDADWIM Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
LYRM9 is a protein coding gene.
Specifications
Specifications
| Antigen | LYRM9 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | LYRM9 |
| Gene Accession No. | A8MSI8 |
| Gene Alias | 1810012P15Rik; C17orf108; C19H17orf108; C9H17orf108; HSD24; LYR motif containing 9; LYR motif-containing protein 9; LYRM9; RGD1562012; UPF0631 protein C17orf108; UPF0631 protein C17orf108 homolog; UPF0631 protein HSD24 homolog; zgc:193694; zgc:193700 |
| Gene Symbols | LYRM9 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?