missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Macrophage Marker Monoclonal Antibody (LN-5)
GREENER_CHOICE

Product Code. 11530872
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11530872 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11530872 Supplier Invitrogen™ Supplier No. MA135492

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LN-5 reacts with human macrophages and displays lymph node germinal center and mantle zone B cell reactivity. It reacts with interdigitating reticulum cells, with tingible body and sinus histiocytes. It further reacts with certain tumor cells and also with normal nonlymphoid tissue like chief cells of the stomach and spermatogonia. LN-5 is negative on Hodgkin's disease and non-Hodgkin's lymphomas.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Macrophage Marker
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LN-5
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias ED2-like antigen; Macrophages; Mature Macrophages
Host Species Mouse
Immunogen Deparaffinized cell extracts of peripheral blood mononuclear cells.
Purification Method purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.