missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MAD1L1/MAD1 Monoclonal specifically detects MAD1L1/MAD1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
Specifications
| Antigen | MAD1L1/MAD1 |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 9B10 |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/Immunofluorescence 10-20 μg/mL, Immunoprecipitation 1:10-1:500 |
| Gene Alias | hMAD1, MAD1 mitotic arrest deficient-like 1 (yeast), MAD1-like protein 1, MAD1yeast, homolog-like 1, Mitotic checkpoint MAD1 protein homolog, Tax-binding protein 181, tumor protein p53 inducible protein 9, TXBP181MAD1 (mitotic arrest deficient, yeast, homolog)-like 1 |
| Gene Symbols | MAD1L1 |
| Host Species | Mouse |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?