missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEC3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21491-1-AP-20UL
This item is not returnable.
View return policy
Description
MAGEC3 is a member of the MAGEC gene family. The members of this family are not expressed in normal tissues, except for testis, and are expressed in tumors of various histological types. The MAGEC genes are clustered on chromosome Xq26-q27. Two transcript variants encoding distinct isoforms have been found for this gene.Specifications
| MAGEC3 | |
| Polyclonal | |
| Unconjugated | |
| MAGEC3 | |
| Cancer/testis antigen 7.2, CT7.2, HCA2, MAGE C3, MAGE C3 antigen, MAGEC3, MAGEC4, melanoma antigen family C, 3, Melanoma associated antigen C3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 139081 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8TD91 | |
| MAGEC3 | |
| MAGEC3 Fusion Protein Ag15819 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction