missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEC3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | MAGEC3 |
|---|---|
| Concentration | 0.2 mg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16831085
|
Proteintech
21491-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16821085
|
Proteintech
21491-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAGEC3 is a member of the MAGEC gene family. The members of this family are not expressed in normal tissues, except for testis, and are expressed in tumors of various histological types. The MAGEC genes are clustered on chromosome Xq26-q27. Two transcript variants encoding distinct isoforms have been found for this gene.Specifications
| MAGEC3 | |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| Q8TD91 | |
| 139081 | |
| MAGEC3 Fusion Protein Ag15819 | |
| Primary | |
| -20°C | |
| MAGEC3 |
| 0.2 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Cancer/testis antigen 7.2, CT7.2, HCA2, MAGE C3, MAGE C3 antigen, MAGEC3, MAGEC4, melanoma antigen family C, 3, Melanoma associated antigen C3 | |
| MAGEC3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title