missing translation for 'onlineSavingsMsg'
Learn More

Mandelamidoxime, 97%, Thermo Scientific™

Product Code. 15484728
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15484728 2 g 2g
15494728 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15484728 Supplier Thermo Scientific Alfa Aesar Supplier No. H58446.04

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24440-15-1
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00194542
InChI Key GXYVISXLFBQBKH-UHFFFAOYSA-N
Synonym mandelamidoxime, 1z-n',2-dihydroxy-2-phenylethanimidamide, n',2-dihydroxy-2-phenylacetimidamide, benzeneethanimidamide, n',alpha-dihydroxy, 2-amino-2-hydroxyimino-1-phenylethan-1-ol, benzeneethanimidamide, n,alpha-dihydroxy-9ci, e-n',2-dihydroxy-2-phenylethanimidamide
PubChem CID 9656867
IUPAC Name N',2-dihydroxy-2-phenylethanimidamide
SMILES C1=CC=C(C=C1)C(C(=NO)N)O

Specifications

Melting Point 148°C to 152°C
Quantity 2 g
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material Mandelamidoxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.