missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2K3 Rabbit anti-Human, Mouse, Rat, CoraLite 488, Clone: 3O10, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech CL488-80137-100UL
This item is not returnable.
View return policy
Description
MAP2K3 antibody apecifically detetects MAP2K3 antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.Specifications
| MAP2K3 | |
| 3O10 | |
| CoraLite 488 | |
| BC032478 | |
| Rabbit | |
| Protein A purification | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunofluorescence | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| MAP2K3 | |
| Peptide | |
| 100 μL | |
| 5606 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction