missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2K3 Rabbit anti-Human, Mouse, Rat, CoraLite 488, Clone: 3O10, Proteintech
Rabbit Recombinant Antibody
145.00€ - 450.00€
Specifications
| Antigen | MAP2K3 |
|---|---|
| Clone | 3O10 |
| Applications | Immunofluorescence |
| Conjugate | CoraLite 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17771329
|
Proteintech
CL488-80137-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17659886
|
Proteintech
CL488-80137-100UL |
100 μL |
450.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAP2K3 antibody apecifically detetects MAP2K3 antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.Specifications
| MAP2K3 | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse, Rat | |
| BC032478 | |
| MAP2K3 | |
| IgG | |
| Protein A purification |
| 3O10 | |
| CoraLite 488 | |
| Rabbit | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 5606 | |
| Peptide | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title