Learn More
MAP2K3 Rabbit, anti-Human, Unconjugated, Clone: 3O10, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand:  Proteintech 80137-1-RR-100UL
 View more versions of this product
Product Code. 17271536
  • 439.00€ / 100µL
Description
MAP2K3, also named as MKK3, is a member of dual specificity kinase group and serves as a specific activator of p38 MAPK with MKK6. MKK3 can be phosphorylated by cytokines and environmental stress at sites Ser189 and Thr193 by MKKK proteins (MEKK 1-4). MKK3 plays a key role in cell differentiation, motility, division and death. It has been reported that MKK3 targeting may represent an effective and potentially safe therapeutic strategy to selectively kill cancer cells in colorectal cancer patients. (PMID: 31695024, 30770795, 26805700, 8622669)
Specifications
Specifications
MAP2K3
Monoclonal
500 μg/mL
Rabbit
Primary
Human, Mouse, Rat
Western Blot, Immunofluorescence, ELISA
3O10
Unconjugated
100 μL
5606