missing translation for 'onlineSavingsMsg'
Learn More

MAP2K3 Rabbit, anti-Human, Unconjugated, Clone: 3O10, Proteintech

Rabbit Recombinant Monoclonal Antibody

149.00€ - 439.00€

Specifications

Antigen MAP2K3
Clone 3O10
Concentration 500 μg/mL
Applications Western Blot, Immunofluorescence, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17281536
View Documents
Proteintech
80137-1-RR-20UL
20 μL
149.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17271536
View Documents
Proteintech
80137-1-RR-100UL
100 μL
439.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MAP2K3, also named as MKK3, is a member of dual specificity kinase group and serves as a specific activator of p38 MAPK with MKK6. MKK3 can be phosphorylated by cytokines and environmental stress at sites Ser189 and Thr193 by MKKK proteins (MEKK 1-4). MKK3 plays a key role in cell differentiation, motility, division and death. It has been reported that MKK3 targeting may represent an effective and potentially safe therapeutic strategy to selectively kill cancer cells in colorectal cancer patients. (PMID: 31695024, 30770795, 26805700, 8622669)

Specifications

Specifications

MAP2K3
500 μg/mL
Monoclonal
Liquid
Human, Mouse, Rat
Primary
3O10
Western Blot, Immunofluorescence, ELISA
Unconjugated
Rabbit
5606
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.