missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAPKAPK3 Recombinant Rabbit Monoclonal Antibody (K01_2N31)
GREENER_CHOICE

Product Code. 30825531
Change view
Click to view available options
Quantity:
50 μL
100 μL
Unit Size:
100µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30825531 100 μL 100µL
30828099 50 μL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30825531 Supplier Invitrogen™ Supplier No. MA564124

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

MAPKAPK3 Recombinant Monoclonal Antibody for Western Blot

MAPKAPK3 functions as an integrative element of signaling in both mitogen and stress responses. MAPKAPK3 functions as a mitogen-activated protein kinase (MAP kinase)- activated protein kinase. MAP kinases are also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals. MAPKAPK3 was shown to be activated by growth inducers and stress stimulation of cells. In vitro studies demonstrated that ERK, p38 MAP kinase and Jun N-terminal kinase were all able to phosphorylate and activate this kinase, which suggested the role of this kinase as an integrative element of signaling in both mitogen and stress responses. MAPKAPK3 was reported to interact with, phosphorylate and repress the activity of E47, which is a basic helix-loop-helix transcription factor known to be involved in the regulation of tissue-specific gene expression and cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAPKAPK3
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2N31
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene MAPKAPK3
Gene Accession No. Q16644
Gene Alias 3PK; AI874665; chromosome 3p kinase; MAP kinase-activated protein kinase 3; MAPK-activated protein kinase 3; MAPKAP kinase 3; MAPKAP3; MAPKAPK3; MAPKAP-K3; MAPKAPK-3; MapkKapk3; mitogen activated protein kinase-activated protein kinase 3; mitogen-activated protein kinase-activated protein kinase 3; MK3; MK-3; OTTHUMP00000210806; OTTHUMP00000210807
Gene Symbols MAPKAPK3
Host Species Rabbit
Immunogen A synthetic peptide of human MK-3.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7867
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.