missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MARK3 Recombinant Rabbit Monoclonal Antibody (24GB3795)

Rabbit Recombinant Monoclonal Antibody
151.00€ - 514.00€
Specifications
| Antigen | MARK3 |
|---|---|
| Clone | 24GB3795 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275122
![]() |
Invitrogen™
MA557063 |
100 μL |
514.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275322
|
Invitrogen™
MA557064 |
20 μL |
151.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MARK3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
MARK3 is a member of a group of kinases known to regulate cell polarity via phosphorylation of microtubule-associated proteins (MAPs). The protein encoded by MARK3 is activated by phosphorylation and in turn is involved in the phosphorylation of tau proteins MAP2 and MAP4. Several transcript variants encoding different isoforms have been found for MARK3.Specifications
| MARK3 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| MARK3 | |
| 1600015G02Rik; A430080F22Rik; cdc25C-associated protein kinase 1; cTAK1; C-TAK1; ELKL motif kinase 2; Emk2; EMK-2; EMK2/MARK3 serine-threonine protein kinase; ETK1; ETK-1; KP78; MAP/microtubule affinity regulating kinase 3; MAP/microtubule affinity-regulating kinase 3; MARK3; microtubule affinity regulating kinase 3; Mpk10; MPK-10; PAR1A; Par-1a; Protein kinase STK10; ser/Thr protein kinase PAR-1; Serine/threonine-protein kinase p78 | |
| MARK3 | |
| Primary | |
| Affinity chromatography |
| 24GB3795 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P27448, Q03141, Q8VHF0 | |
| 170577, 17169, 4140 | |
| A synthesized peptide derived from human MARK3 (aa 550-600). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title