missing translation for 'onlineSavingsMsg'
Learn More

MART-1 / MLANA Rabbit, Clone: MLANA/1409R, enQuireBio™

Product Code. p-7156046
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15920018 20 μg 20µg
15930018 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15920018 Supplier EnquireBio Supplier No. 2315RBM9P0

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Specifications

Antigen MART-1 / MLANA
Applications Immunohistochemistry
Cellular Localization Cytoplasmic
Classification Monoclonal
Clone MLANA/1409R
Concentration 200μg/mL
Conjugate Unconjugated
Formulation 10mM PBS with or without 0.05% BSA and 0.05% azide.
Gene Accession No. NP_005502
Gene Symbols MLANA
Host Species Rabbit
Immunogen Recombinant human full-length MLANA protein
Molecular Weight of Antigen 20-22kDa (doublet)
Positive Control SK-MEL-13 and SK-MEL-19 Melanoma cell lines; Melanomas
Purification Method Protein A/G Purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2315
Content And Storage Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.