missing translation for 'onlineSavingsMsg'
Learn More

MCM7 Mouse, Clone: MCM7/1468, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 4176-MSM3-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15920458

  • 510.00€ / 100µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MCM7
Nuclear
MCM7/1468
Unconjugated
NP_005907
Mouse
88kDa
Protein A/G Purified
RUO
4176
IgG2b
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
MCM7
Recombinant human MCM7 protein fragment (aa195-319)
Raji or Ramos cells. Tonsil
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions