Learn More
MECP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10861-1-AP-20UL
 View more versions of this product
Product Code. 16837533
  • 145.00€ / 20µL
Description
This antibody reacts with the MECP2 and phosphorylated MECP2 proteins. The calculated molecular weight of MECP2 is 52 kDa, but the post-modified MECP2 protein is about 75-80 kDa.
MECP2 belongs to a family of nuclear proteins (including MBD1, MBD2, MBD3 and MBD4) that have a methyl-CpG binding domain. MECP2 is capable of binding specifically to methylated DNA. DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. MECP2 can also repress transcription from methylated gene promoters. In contrast to other MBD family members, MECP2 is X-linked and subject to X inactivation. MECP2 is dispensible in stem cells, but is essential for embryonic development. MECP2 gene mutations are the cause of most cases of Rett syndrome, a progressive neurologic developmental disorder and one of the most common causes of mental retardation in females.
Specifications
Specifications
MECP2
Polyclonal
Unconjugated
MECP2
AUTSX3, MeCp 2 protein, MECP2, Methyl CpG binding protein 2, MRX16, MRX79, MRXS13, MRXSL, PPMX, RTS, RTT
Rabbit
Antigen Affinity Chromatography
RUO
17257, 29386, 4204
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Flow Cytometry
0.18 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P51608, Q00566, Q9Z2D6
Mecp2
MECP2 Fusion Protein Ag1309
20 μL
Primary
Mouse, Rat, Human