missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MECP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | MECP2 |
|---|---|
| Concentration | 0.18 mg/mL |
| Applications | Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16837533
|
Proteintech
10861-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16827533
|
Proteintech
10861-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody reacts with the MECP2 and phosphorylated MECP2 proteins. The calculated molecular weight of MECP2 is 52 kDa, but the post-modified MECP2 protein is about 75-80 kDa.
MECP2 belongs to a family of nuclear proteins (including MBD1, MBD2, MBD3 and MBD4) that have a methyl-CpG binding domain. MECP2 is capable of binding specifically to methylated DNA. DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. MECP2 can also repress transcription from methylated gene promoters. In contrast to other MBD family members, MECP2 is X-linked and subject to X inactivation. MECP2 is dispensible in stem cells, but is essential for embryonic development. MECP2 gene mutations are the cause of most cases of Rett syndrome, a progressive neurologic developmental disorder and one of the most common causes of mental retardation in females.Specifications
| MECP2 | |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| P51608, Q00566, Q9Z2D6 | |
| 17257, 29386, 4204 | |
| MECP2 Fusion Protein Ag1309 | |
| Primary | |
| -20°C | |
| MECP2 |
| 0.18 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| AUTSX3, MeCp 2 protein, MECP2, Methyl CpG binding protein 2, MRX16, MRX79, MRXS13, MRXSL, PPMX, RTS, RTT | |
| Mecp2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title