missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES, 0.5M buffer soln., pH 6.5

Product Code. 15435949
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435949 100 mL 100mL
15445949 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435949 Supplier Thermo Scientific Chemicals Supplier No. J63778.AE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 145224-94-8 | C6H15NO5S | 213.248 g/mol

MES is used as a running buffer for resolving very small proteins (ex: 13 kDa) on bis-tris gels. Also used in cation-exchange chromatography and as a biological buffer in plant cell cultures. Used in various protein characterization assays. MES is also useful as a non-coordinating buffer in chemistry involving metal ions, as many common buffers (e.g. phosphate and acetate) readily form coordination complexes. MES only weakly binds Ca, Mg, Mn, and it has negligible binding with Cu(II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES is used as a running buffer for resolving very small proteins (ex: 13 kDa) on bis-tris gels. Also used in cation-exchange chromatography and as a biological buffer in plant cell cultures. Used in various protein characterization assays. MES is also useful as a non-coordinating buffer in chemistry involving metal ions, as many common buffers (e.g. phosphate and acetate) readily form coordination complexes. MES only weakly binds Ca, Mg, Mn, and it has negligible binding with Cu(II).

Solubility
Soluble in water (∼1.3 M).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.248
MDL Number MFCD00283848
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
PubChem CID 16218417
IUPAC Name 2-morpholin-4-ylethanesulfonic acid;hydrate
SMILES C1COCCN1CCS(=O)(=O)O.O

Specifications

Color Colorless
Concentration 0.5 M
pH 6.5
Physical Form Liquid
Quantity 100 mL
Solubility Information Soluble in water (∽1.3 M).
Chemical Name or Material MES buffer soln.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.