missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES-buffered saline (5X), pH 6.5

Product Code. 15414459
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15414459 250 mL 250mL
15424459 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15414459 Supplier Thermo Scientific Chemicals Supplier No. J61979.AK
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 145224-94-8 | C6H15NO5S | 213.248 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00149409
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
PubChem CID 16218417
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate
SMILES O.OS(=O)(=O)CCN1CCOCC1

Specifications

Concentration 5X
pH 6.5
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material MES-buffered saline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.