missing translation for 'onlineSavingsMsg'
Learn More

Methoxyacetic acid, 97%

Product Code. 11419977
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419977 50 g 50g
11429977 250 g 250g
11409977 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419977 Supplier Thermo Scientific Alfa Aesar Supplier No. B22917.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 625-45-6 | C3H6O3 | 90.078 g/mol

Methoxyacetic acid is used in zebrafish embryotoxicity test to evaluate chemical toxicity after exposure of the complete vertebrate embryo during embryogenesis. It is also used in industry as gelling, viscosity and stabilizer reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methoxyacetic acid is used in zebrafish embryotoxicity test to evaluate chemical toxicity after exposure of the complete vertebrate embryo during embryogenesis. It is also used in industry as gelling, viscosity and stabilizer reagent.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-45-6
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.078
MDL Number MFCD00004308
InChI Key RMIODHQZRUFFFF-UHFFFAOYSA-N
Synonym methoxyacetic acid, acetic acid, methoxy, methoxyethanoic acid, acetic acid, 2-methoxy, ch3och2cooh, methoxyacetlc acid, unii-f11t1h7q7w, methoxy acetic acid, methoxy-acetic acid, ccris 6518
PubChem CID 12251
IUPAC Name 2-methoxyacetic acid
SMILES COCC(=O)O

Specifications

Melting Point 8°C
Density 1.174
pH 1.6
Boiling Point 202°C to 204°C
Flash Point 114°C (237°F)
Odor Odorless
Linear Formula CH3OCH2CO2H
Refractive Index 1.4165
Quantity 50 g
UN Number UN3265
Beilstein 635800
Solubility Information Miscible with water.
Formula Weight 90.08
Percent Purity 97%
Chemical Name or Material Methoxyacetic acid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methoxyacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Methoxyacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.