missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-aminobenzoate, 98%

Product Code. 11487826
Click to view available options
Quantity:
5 g
25 g
100 g
missing translation for 'unitSize'
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11487826

missing translation for 'mfr': Thermo Scientific Alfa Aesar A14766.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4518-10-9 | C8H9NO2 | 151.17 g/mol

Methyl 3-aminobenzoate, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-aminobenzoate, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water (0.1-1%) .

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4518-10-9
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00017102
InChI Key VZDNXXPBYLGWOS-UHFFFAOYSA-N
Synonym benzoic acid, 3-amino-, methyl ester, methyl-3-aminobenzoate, 3-aminobenzoic acid methyl ester, m-aminobenzoic acid, methyl ester, 3-amino-benzoic acid methyl ester, methyl m-aminobenzoate, methyl3-aminobenzoate, methyl-m-aminobenzoate, pubchem16709, acmc-1aeif
PubChem CID 78274
IUPAC Name methyl 3-aminobenzoate
SMILES COC(=O)C1=CC(=CC=C1)N

Specifications

Melting Point 49°C to 53°C
Density 1.232
Quantity 25 g
Beilstein 1101468
Solubility Information Slightly soluble in water (0.1-1%) .
Formula Weight 151.17
Percent Purity ≥98%
Chemical Name or Material Methyl 3-aminobenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.