missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(methylsulfonylmethyl)benzoate, 98%

Product Code. 11317535
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317535 2 g 2g
11327535 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317535 Supplier Thermo Scientific Alfa Aesar Supplier No. L05657.04
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 160446-22-0 | C10H12O4S | 228.262 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 160446-22-0
Molecular Formula C10H12O4S
Molecular Weight (g/mol) 228.262
MDL Number MFCD00051693
InChI Key XCKJFFWCTKJUKD-UHFFFAOYSA-N
Synonym methyl 4-methylsulfonylmethyl benzoate, methyl 4-methanesulfonylmethyl benzoate, methyl 4-methylsulfonyl methyl benzoate, benzoic acid, 4-methylsulfonyl methyl-, methyl ester, acmc-20akgt, maybridge1_002811, methyl 4-methanesulphonylmethyl benzoate
PubChem CID 2804364
IUPAC Name methyl 4-(methylsulfonylmethyl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)CS(=O)(=O)C

Specifications

Melting Point 164°C to 166°C
Quantity 2 g
Beilstein 7810971
Formula Weight 228.27
Percent Purity 98%
Chemical Name or Material Methyl 4-(methylsulfonylmethyl)benzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.