missing translation for 'onlineSavingsMsg'
Learn More

Methyl Tricosanoate, TRC

Product Code. 17947524
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17947524 100 mg 100mg
17940065 500 mg 500mg
17989634 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17947524 Supplier Toronto Research Chemicals Supplier No. TRCM331185100MG
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2433-97-8
Molecular Formula C24 H48 O2
Molecular Weight (g/mol) 368.64
Synonym Tricosanoic acid methyl ester
IUPAC Name methyl tricosanoate
SMILES CCCCCCCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C24H48O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24(25)26-2/h3-23H2,1-2H3
Formula Weight 368.3654
Percent Purity >95%
Chemical Name or Material Tricosanoic acid methyl ester
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.