missing translation for 'onlineSavingsMsg'
Learn More

Methylene Violet 3RAX (~90%), TRC

Product Code. 30466293
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466293 5 g 5g
30476809 1 g 1g
30474977 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30466293 Supplier Toronto Research Chemicals Supplier No. TRCM2367555G
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4569-86-2
Molecular Formula C22H23N4 . Cl
Molecular Weight (g/mol) 378.9
Synonym 3-Amino-7-(diethylamino)-5-phenylphenazin-5-ium Chloride, C.I. 50206, Diethylsafranin, 3-Amino-7-diethylamino-5-phenylphenazinium Chloride
IUPAC Name 3-amino-7-(diethylamino)-5-phenylphenazin-5-ium chloride
SMILES CCN(C1=CC2=[N+](C3=CC=CC=C3)C4=CC(N)=CC=C4N=C2C=C1)CC.[Cl-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C22H22N4.ClH/c1-3-25(4-2)18-11-13-20-22(15-18)26(17-8-6-5-7-9-17)21-14-16(23)10-12-19(21)24-20;/h5-15,23H,3-4H2,1-2H3;1H
Formula Weight 378.16
Chemical Name or Material Methylene Violet 3RAX
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.