Learn More
METTL5 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Product Code. 18085385 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18085385 100 μL 100µL
18035555 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18085385 Supplier Proteintech Supplier No. CL48816791100UL
Rabbit Polyclonal Antibody
The METTL5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human METTL5. This antibody recognizes human antigen. The METTL5 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Antigen METTL5
Applications Immunofluorescence
Concentration 1000 μg/mL
Conjugate CoraLite Plus 488
Gene Symbols FLJ10459, HSPC133, methyltransferase like 5, METTL5
Host Species Rabbit
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 29081
Target Species Human
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG