Learn More
Mettler Toledo™ DL12K Buret Tubing for Mettler Toledo Titrators
Product Code. 10773212 Shop All Mettler Toledo Products
Change view
Click to view available options
Type:
Suction Tube
Length (English):
32.62 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Length (English) Unit Size
10773212 Suction Tube 32.62 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10773212 Supplier Mettler Toledo™ Supplier No. 00025688
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
For use with Mettler Toledo titrators.
Use the 09-313-151J suction tube and 09-313-151H dispensing tubing with siphon tip as spares for DV1000 Series Burettes.
TRUSTED_SUSTAINABILITY
Specifications
Type Suction Tube
Length (English) 32.62 in.
Length (Metric) 830 mm