Description
- Type: SC-Lemo
- For use with standard and Plug and Play electrodes
- For V10/20S (01-914-315/ 01-914-315) or G10/20S (01-914-318/ 01-914-319) series titrators
- Length: 100cm
Specifications
Specifications
| Connector Types | Lemo |
| For Use With (Equipment) | Standard and Plug and Play Electrodes |
| For Use With (Application) | Connection to the titrators |
| Includes | S12 grey connection |
| Length (Metric) Cable | 100 cm |