missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MGST3 Recombinant Rabbit Monoclonal Antibody (K01_1T87)
GREENER_CHOICE

Product Code. 30824819
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30824819 100 μL
30827307 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30824819 Supplier Invitrogen™ Supplier No. MA563741

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

MGST3 Recombinant Monoclonal Antibody for IHC (P), Western Blot

The MAPEG family consists of six human proteins, several of which are involved the production of leukotrienes and prostaglandin E, important mediators of inflammation. This gene encodes an enzyme which catalyzes the conjugation of leukotriene A4 and reduced glutathione to produce leukotriene C4. This enzyme also demonstrates glutathione-dependent peroxidase activity towards lipid hydroperoxides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGST3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_1T87
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene MGST3
Gene Accession No. O14880
Gene Alias 2010012L10Rik; 2010306B17Rik; 2700004G04Rik; AA516734; Glutathione peroxidase MGST3; GST-III; LTC4 synthase MGST3; MGST3; microsomal glutathione S-transferase; microsomal glutathione S-transferase 3; Microsomal glutathione S-transferase III; microsomal GST-3; Microsomal GST-III
Gene Symbols MGST3
Host Species Rabbit
Immunogen A synthetic peptide of human MGST3.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4259
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.