missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class I Antibody (ER-HR52) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-64952
This item is not returnable.
View return policy
Description
MHC Class I Monoclonal specifically detects MHC Class I in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| MHC Class I | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 1:50-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence | |
| HLA-E | |
| Adherent FI (CBAXBL) bone marrow stromal cells | |
| 0.125 mg | |
| Adaptive Immunity, Immunology, Major Histocompatibility Complexes | |
| 3133 | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| ER-HR52 | |
| Unconjugated | |
| DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-64952 recognizes a polymorphic determinant on the murine MHC (major histocompatibility complex) class I molecule. In the mouse, the MHC class I molecule is expressed on the cell surface of all nucleated cells but expression levels may differ according to cell type. Clone ER-MP52 specifically recognizes mouse strains with the haplotypes H-2b,w16 and H-2d,p,q. Mouse strains with the haplotypes H-2f,r,s,w17,w23,w27 show weak reactivity with this antibody. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction