missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class I Antibody (ER-HR52) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 1 publication
204.00€
Specifications
| Antigen | MHC Class I |
|---|---|
| Clone | ER-HR52 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 1:50-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18790954
|
Novus Biologicals
NB100-64952SS |
0.025 mg |
204.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18009265
|
Novus Biologicals
NB100-64952 |
0.125 mg |
N/A
|
N/A | |||||
Description
MHC Class I Monoclonal specifically detects MHC Class I in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| MHC Class I | |
| 1.0 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rat | |
| Adaptive Immunity, Immunology, Major Histocompatibility Complexes | |
| DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 | |
| HLA-E | |
| IgG2a | |
| Protein G purified | |
| NB100-64952 recognizes a polymorphic determinant on the murine MHC (major histocompatibility complex) class I molecule. In the mouse, the MHC class I molecule is expressed on the cell surface of all nucleated cells but expression levels may differ according to cell type. Clone ER-MP52 specifically recognizes mouse strains with the haplotypes H-2b,w16 and H-2d,p,q. Mouse strains with the haplotypes H-2f,r,s,w17,w23,w27 show weak reactivity with this antibody. |
| ER-HR52 | |
| Flow Cytometry 1:50-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| 3133 | |
| Adherent FI (CBAXBL) bone marrow stromal cells | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title