Learn More
MHC Class I Antibody (ER-HR52) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NB100-64952SS
 View more versions of this product
Product Code. 18790954
  • 204.00€ / 25µg
Description
MHC Class I Monoclonal specifically detects MHC Class I in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MHC Class I
Monoclonal
1.0 mg/mL
Flow Cytometry 1:50-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence
HLA-E
Adherent FI (CBAXBL) bone marrow stromal cells
0.025 mg
Adaptive Immunity, Immunology, Major Histocompatibility Complexes
3133
Mouse
Purified
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
ER-HR52
Unconjugated
DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1
Rat
Protein G purified
RUO
Primary
NB100-64952 recognizes a polymorphic determinant on the murine MHC (major histocompatibility complex) class I molecule. In the mouse, the MHC class I molecule is expressed on the cell surface of all nucleated cells but expression levels may differ according to cell type. Clone ER-MP52 specifically recognizes mouse strains with the haplotypes H-2b,w16 and H-2d,p,q. Mouse strains with the haplotypes H-2f,r,s,w17,w23,w27 show weak reactivity with this antibody.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.